Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037906.2 | NP_001032995.1 | 1 - 2 | 97 | 124 | |
GenBank mRNA | AK032845.1 | - | 1 - 2 | 379 | 124 | |
AY622226.1 | - | 1 - 2 | 97 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |