Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114116.1 | NP_001107588.1 | 2 - 3 | 196 | 74 | |
GenBank mRNA | AU080420.1 | - | 2 - 3 | 196 | 74 | |
BC051969.1 | - | 1 - 2 | 104 | 5694653 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |