Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005255.1 | - | 3 - 4 | 467 | 90 | |
AK188228.1 | - | 2 - 3 | 254 | 90 | ||
AK205856.1 | - | 2 - 3 | 254 | 90 | ||
AK214551.1 | - | 2 - 3 | 254 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |