Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hectd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163471.1 | NP_001156943.1 | 6 - 7 | 807 | 154 | |
NM_172637.3 | NP_766225.2 | 6 - 7 | 807 | 154 | ||
GenBank mRNA | AK029549.1 | - | 6 - 7 | 667 | 154 | |
AK043646.1 | - | 6 - 7 | 768 | 154 | ||
BC137671.1 | - | 6 - 7 | 778 | 8860690 | 154 | |
BC144815.1 | - | 6 - 7 | 778 | 9053363 | 154 | |
CB526486.1 | - | 3 - 4 | 260 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |