Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900010333.mm.1 |
TC1900001104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010334.mm.1 |
TC1900001104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010339.mm.1 |
NM_025333.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010339.mm.1 |
NM_025333.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010339.mm.1 |
TC1900001104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010341.mm.1 |
NM_025333.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010341.mm.1 |
NM_025333.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010341.mm.1 |
TC1900001104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010337.mm.1 |
NM_025333.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010337.mm.1 |
NM_025333.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010337.mm.1 |
TC1900001104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010336.mm.1 |
NM_025333.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010336.mm.1 |
NM_025333.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010336.mm.1 |
TC1900001104.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010340.mm.1 |
NM_025333.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010340.mm.1 |
NM_025333.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010340.mm.1 |
TC1900001104.mm.1 |
MTA TC |