Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198163.1 | NP_937806.1 | 3 - 4 | 386 | 70 | |
GenBank mRNA | AB232631.1 | - | 3 - 4 | 226 | 70 | |
AK156576.1 | - | 3 - 4 | 377 | 70 | ||
BC030941.1 | - | 1 - 2 | 77 | 70 | ||
BC056466.1 | - | 3 - 4 | 386 | 5717246 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |