Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009009.4 | NP_033035.3 | 8 - 9 | 1188 | 67 | |
GenBank mRNA | AF332085.1 | - | 7 - 8 | 943 | 67 | |
AF332086.1 | - | 7 - 8 | 943 | 67 | ||
AK004746.1 | - | 8 - 9 | 991 | 67 | ||
AK128984.1 | - | 8 - 9 | 1077 | 67 | ||
AK129050.1 | - | 3 - 4 | 316 | 67 | ||
AK139492.1 | - | 7 - 8 | 841 | 67 | ||
AK145591.1 | - | 8 - 9 | 1155 | 67 | ||
AK163992.1 | - | 1049 | 67 | |||
AK168718.1 | - | 8 - 9 | 1176 | 67 | ||
AK168896.1 | - | 8 - 9 | 1137 | 67 | ||
BC043032.1 | - | 7 - 8 | 957 | 5696540 | 67 | |
BC129916.1 | - | 7 - 8 | 987 | 40111820 | 67 | |
BC129917.1 | - | 7 - 8 | 988 | 40111821 | 67 | |
X98293.1 | - | 7 - 8 | 943 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |