Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010360.2 | NP_034490.1 | 2 - 3 | 226 | 73 | |
GenBank mRNA | AK002750.1 | - | 2 - 3 | 158 | 73 | |
AK019317.1 | - | 2 - 3 | 157 | 73 | ||
AK028098.1 | - | 2 - 3 | 242 | 73 | ||
BC008206.1 | - | 2 - 3 | 180 | 3154170 | 73 | |
U24428.1 | - | 2 - 3 | 222 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |