Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hells | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008234.3 | NP_032260.2 | 20 - 21 | 2390 | 118 | |
GenBank mRNA | AF155210.1 | - | 20 - 21 | 2390 | 118 | |
AK013266.1 | - | 20 - 21 | 2429 | 118 | ||
AK147126.1 | - | 20 - 21 | 2421 | 118 | ||
AK166636.1 | - | 20 - 21 | 2391 | 118 | ||
BC020056.1 | - | 7 - 8 | 754 | 118 | ||
BC090671.1 | - | 20 - 21 | 2433 | 117 | ||
BC100394.1 | - | 20 - 21 | 2387 | 30480539 | 118 | |
U25691.2 | - | 20 - 21 | 2423 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |