Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851465 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
307371 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
130378 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
903628 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
66821 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355450 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
601902 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
59811 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
866834 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364157 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
788283 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
96920 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
135853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
753618 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
755484 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
272134 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676754 |
AF148857.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676754 |
AK142886.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676754 |
NM_013788.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324318 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
104724 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
58265 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
87204 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
23544 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140663 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P146259 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104669 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104694 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104663 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104679 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104676 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104660 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104662 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104659 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104667 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104680 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104657 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104691 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104684 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104692 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104671 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104678 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104665 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104690 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104687 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104683 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104696 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32104673 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P140663 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P146259 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09168952 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09168901 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09168924 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P447785 |
AK008580.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P447785 |
AY206875.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P447785 |
BC010824.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P447785 |
NM_028089.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125829 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P146259 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P140663 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14199596 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14430492 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14430495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32104763 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14199601 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104653 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104721 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104700 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104694 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104676 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104645 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104662 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104692 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104714 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14201100 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104690 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14252534 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104702 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104639 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104696 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14252535 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104665 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104635 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14295984 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104657 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104671 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14295987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104719 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104683 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104685 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104716 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14199602 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104655 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104680 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104674 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104640 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104708 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104687 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14253368 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104669 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104720 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14430493 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14505770 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104663 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104637 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14306054 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14499544 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104633 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14444691 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104641 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104684 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104654 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32104723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14499547 |
- |
- |