Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102471.1 | NP_001095941.1 | 1 - 2 | 1784 | 65 | |
NM_033569.3 | NP_291047.2 | 1 - 2 | 1784 | 65 | ||
GenBank mRNA | AF216961.1 | - | 1 - 2 | 1096 | 65 | |
AK032006.1 | - | 1 - 2 | 1743 | 65 | ||
AK044400.1 | - | 1 - 2 | 1788 | 65 | ||
AK053711.1 | - | 1 - 2 | 1802 | 65 | ||
AK159616.1 | - | 1 - 2 | 1790 | 65 | ||
BC012956.1 | - | 1 - 2 | 1780 | 65 | ||
BC027387.1 | - | 1 - 2 | 1581 | 65 | ||
BC125033.1 | - | 1 - 2 | 1096 | 65 | ||
GQ225745.1 | - | 1 - 2 | 1083 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |