Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033174.1 | NP_001028346.1 | 11 - 12 | 1971 | 144 | |
GenBank mRNA | AK132663.1 | - | 9 - 10 | 1251 | 144 | |
AK220301.1 | - | 8 - 9 | 1009 | 144 | ||
BC094342.1 | - | 5 - 6 | 592 | 144 | ||
BC107327.1 | - | 11 - 12 | 1705 | 144 | ||
BC107328.2 | - | 11 - 12 | 1705 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |