Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027156.3 | NP_081432.2 | 8 - 9 | 96 | ||
GenBank mRNA | AK141361.1 | - | 8 - 9 | 96 | ||
AK152599.1 | - | 8 - 9 | 96 | |||
BC023700.1 | - | 5 - 6 | 462 | 96 | ||
BC060646.1 | - | 8 - 9 | 6853826 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |