Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042655.1 | NP_001036120.1 | 5 - 6 | 730 | 133 | |
GenBank mRNA | AK039566.1 | - | 5 - 6 | 545 | 133 | |
AK046883.1 | - | 5 - 6 | 564 | 133 | ||
AK081318.1 | - | 5 - 6 | 543 | 133 | ||
AK083153.1 | - | 5 - 6 | 548 | 133 | ||
AK087754.1 | - | 555 | 133 | |||
AK136705.1 | - | 5 - 6 | 555 | 133 | ||
AK149917.1 | - | 5 - 6 | 542 | 133 | ||
AK153732.1 | - | 5 - 6 | 542 | 133 | ||
AK170821.1 | - | 5 - 6 | 542 | 133 | ||
BC017607.1 | - | 5 - 6 | 730 | 4507589 | 133 | |
BC035449.1 | - | 5 - 6 | 529 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |