Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163662.1 | NP_001157134.1 | 6 - 7 | 758 | 60 | |
NM_008749.2 | NP_032775.1 | 6 - 7 | 758 | 60 | ||
GenBank mRNA | AK002630.1 | - | 6 - 7 | 739 | 60 | |
AK004886.1 | - | 6 - 7 | 729 | 60 | ||
AK007976.1 | - | 6 - 7 | 732 | 60 | ||
AK045087.1 | - | 6 - 7 | 758 | 60 | ||
AK140710.1 | - | 6 - 7 | 736 | 60 | ||
AK160981.1 | - | 6 - 7 | 745 | 60 | ||
AK164088.1 | - | 6 - 7 | 740 | 60 | ||
AK170717.1 | - | 6 - 7 | 753 | 60 | ||
AK194015.1 | - | 1 - 2 | 26 | 60 | ||
BC072554.1 | - | 6 - 7 | 736 | 5684979 | 60 | |
M96823.1 | - | 5 - 6 | 680 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |