Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700027366.mm.1 |
NM_001271993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027366.mm.1 |
NM_018876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027366.mm.1 |
TC0700003015.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027364.mm.1 |
NM_001271993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027364.mm.1 |
TC0700003015.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027363.mm.1 |
NM_001271993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027363.mm.1 |
NM_018876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027363.mm.1 |
TC0700003015.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027365.mm.1 |
NM_001271993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027365.mm.1 |
NM_018876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027365.mm.1 |
TC0700003015.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027367.mm.1 |
NM_001271993.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027367.mm.1 |
NM_018876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027367.mm.1 |
TC0700003015.mm.1 |
MTA TC |