Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253814.1 | NP_001240743.1 | 11 - 12 | 2778 | 86 | |
NM_027922.2 | NP_082198.1 | 11 - 12 | 2775 | 86 | ||
GenBank mRNA | AK003234.1 | - | 8 - 9 | 1577 | 86 | |
AK129192.1 | - | 11 - 12 | 1887 | 86 | ||
AK132187.1 | - | 11 - 12 | 2780 | 86 | ||
AK134966.1 | - | 1 - 2 | 610 | 86 | ||
AK136587.1 | - | 1 - 2 | 198 | 86 | ||
AK142497.1 | - | 9 - 10 | 1827 | 86 | ||
BC065071.1 | - | 11 - 12 | 2738 | 6410033 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |