Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133712.2 | NP_598473.1 | 1 - 2 | 97 | 81 | |
GenBank mRNA | AK009000.1 | - | 1 - 2 | 84 | 81 | |
BC002100.1 | - | 1 - 2 | 96 | 3484608 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |