Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173365.2 | NP_775541.1 | 1 - 2 | 183 | 66 | |
GenBank mRNA | AK040547.1 | - | 1 - 2 | 184 | 66 | |
BC117783.2 | - | 1 - 2 | 45 | 40092101 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |