Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033311.3 | NP_001028483.2 | 4 - 5 | 898 | 79 | |
GenBank mRNA | AK138152.1 | - | 4 - 5 | 847 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |