Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133711.3 | NP_598472.2 | 3 - 4 | 477 | 114 | |
GenBank mRNA | AF395083.2 | - | 3 - 4 | 477 | 114 | |
AF530515.1 | - | 3 - 4 | 477 | 114 | ||
AK005612.1 | - | 3 - 4 | 477 | 114 | ||
BC048650.1 | - | 3 - 4 | 477 | 6772684 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |