Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011714.2 | NP_035844.2 | 6 - 7 | 1222 | 88 | |
GenBank mRNA | AF084480.1 | - | 6 - 7 | 1181 | 88 | |
AK017894.1 | - | 6 - 7 | 1223 | 88 | ||
AK137139.1 | - | 2 - 3 | 959 | 88 | ||
AK140172.1 | - | 6 - 7 | 1010 | 88 | ||
AK141305.1 | - | 6 - 7 | 963 | 88 | ||
BC141399.1 | - | 6 - 7 | 1125 | 9056012 | 88 | |
BC171975.1 | - | 6 - 7 | 1125 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |