Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009580.2 | NP_033606.2 | 9 - 10 | 1568 | 63 | |
GenBank mRNA | BC100347.1 | - | 9 - 10 | 1586 | 63 | |
BC125613.1 | - | 9 - 10 | 1568 | 40130128 | 63 | |
BC125615.1 | - | 9 - 10 | 1568 | 40130130 | 63 | |
U20448.1 | - | 9 - 10 | 1599 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |