Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019699.1 | NP_062673.1 | 6 - 7 | 882 | 97 | |
GenBank mRNA | AF126798.1 | - | 6 - 7 | 882 | 97 | |
AK083282.1 | - | 849 | 97 | |||
AK143524.1 | - | 900 | 97 | |||
BC057189.1 | - | 6 - 7 | 870 | 5137343 | 97 | |
HQ264057.1 | - | 6 - 7 | 775 | 97 | ||
HQ264058.1 | - | 6 - 7 | 775 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |