Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029415.2 | NP_083691.1 | 1 - 2 | 548 | 70 | |
GenBank mRNA | AJ583504.1 | - | 1 - 2 | 377 | 70 | |
AK018423.1 | - | 1 - 2 | 549 | 70 | ||
BC127608.1 | - | 1 - 2 | 441 | 40058121 | 70 | |
BC139343.1 | - | 1 - 2 | 440 | 8862365 | 70 | |
BC139346.1 | - | 1 - 2 | 440 | 8862368 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |