Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ibsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008318.3 | NP_032344.2 | 3 - 4 | 183 | 64 | |
GenBank mRNA | AK132371.1 | - | 3 - 4 | 182 | 64 | |
AK162640.1 | - | 3 - 4 | 166 | 64 | ||
BB618282.1 | - | 3 - 4 | 300 | 64 | ||
BC045143.1 | - | 3 - 4 | 168 | 6391115 | 64 | |
L20232.1 | - | 3 - 4 | 165 | 64 | ||
L23801.1 | - | 3 - 4 | 184 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |