Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019457.2 | NP_062330.1 | 3 - 4 | 378 | 166 | |
GenBank mRNA | AF092208.1 | - | 3 - 4 | 323 | 166 | |
AK016236.1 | - | 3 - 4 | 367 | 166 | ||
AK077124.1 | - | 3 - 4 | 362 | 166 | ||
BC046277.1 | - | 3 - 4 | 329 | 6310345 | 166 | |
CJ046721.1 | - | 3 - 4 | 379 | 166 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |