Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd1lg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021396.2 | NP_067371.1 | 1 - 2 | 217 | 69 | |
GenBank mRNA | AF142780.2 | - | 1 - 2 | 190 | 69 | |
AK089369.1 | - | 1 - 2 | 220 | 69 | ||
AK155002.1 | - | 1 - 2 | 157 | 69 | ||
BC104138.1 | - | 1 - 2 | 122 | 40045578 | 69 | |
BC104139.1 | - | 1 - 2 | 122 | 40045580 | 69 | |
BY749628.1 | - | 1 - 2 | 220 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |