Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021299.1 | NP_067274.1 | 1 - 2 | 225 | 59 | |
GenBank mRNA | AB020203.1 | - | 1 - 2 | 128 | 59 | |
AK004864.2 | - | 1 - 2 | 240 | 59 | ||
AK005194.2 | - | 1 - 2 | 279 | 59 | ||
AK007618.1 | - | 276 | 59 | |||
AK008681.1 | - | 1 - 2 | 242 | 59 | ||
AK031631.1 | - | 1 - 2 | 223 | 59 | ||
AK053644.1 | - | 1 - 2 | 285 | 59 | ||
AK089027.1 | - | 1 - 2 | 276 | 59 | ||
AK147060.1 | - | 1 - 2 | 285 | 59 | ||
AK149996.1 | - | 1 - 2 | 282 | 59 | ||
BC016432.1 | - | 1 - 2 | 227 | 4217820 | 59 | |
BC019174.1 | - | 1 - 2 | 278 | 5007578 | 59 | |
BC024871.1 | - | 1 - 2 | 225 | 3154594 | 59 | |
BC058191.1 | - | 1 - 2 | 255 | 5340253 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |