Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021535.4 | NP_067510.3 | 6 - 7 | 832 | 91 | |
GenBank mRNA | AB044414.1 | - | 6 - 7 | 761 | 91 | |
AK003493.1 | - | 6 - 7 | 819 | 91 | ||
AK011140.1 | - | 6 - 7 | 830 | 90 | ||
AK019174.1 | - | 6 - 7 | 791 | 91 | ||
AK145982.1 | - | 6 - 7 | 831 | 91 | ||
AK159635.1 | - | 6 - 7 | 819 | 91 | ||
AK168813.1 | - | 6 - 7 | 826 | 91 | ||
BC020034.1 | - | 6 - 7 | 774 | 91 | ||
BC057446.1 | - | 6 - 7 | 793 | 3155760 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |