Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290448.1 | NP_001277377.1 | 5 - 6 | 1490 | 99 | |
NM_001290449.1 | NP_001277378.1 | 5 - 6 | 1490 | 99 | ||
NM_023739.3 | NP_076228.2 | 5 - 6 | 1453 | 99 | ||
GenBank mRNA | AF223576.1 | - | 5 - 6 | 1417 | 99 | |
AK005038.2 | - | 5 - 6 | 1425 | 99 | ||
AK033850.1 | - | 5 - 6 | 1384 | 99 | ||
AK050749.1 | - | 5 - 6 | 1427 | 99 | ||
AK144647.1 | - | 5 - 6 | 1439 | 99 | ||
AK146108.1 | - | 5 - 6 | 1407 | 99 | ||
AK160013.1 | - | 5 - 6 | 1423 | 99 | ||
BC053709.1 | - | 5 - 6 | 1388 | 30052746 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |