Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009160.2 | NP_033186.1 | 1 - 2 | 74 | 90 | |
GenBank mRNA | BY116790.1 | - | 1 - 2 | 76 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |