Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177816.3 | NP_808484.2 | 1 - 2 | 187 | 79 | |
GenBank mRNA | AX776199.1 | - | 1 - 2 | 187 | 79 | |
BC138964.1 | - | 1 - 2 | 285 | 8861986 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |