Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dydc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027717.1 | NP_081993.1 | 2 - 3 | 230 | 93 | |
GenBank mRNA | AK016963.1 | - | 2 - 3 | 365 | 93 | |
BC113750.1 | - | 2 - 3 | 230 | 40049377 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |