Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wapal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004436.4 | NP_001004436.2 | 11 - 12 | 3069 | 133 | |
NM_001301330.1 | NP_001288259.1 | 11 - 12 | 3051 | 133 | ||
GenBank mRNA | AB167349.1 | - | 11 - 12 | 2875 | 133 | |
AK028167.1 | - | 9 - 10 | 1173 | 133 | ||
AK129105.1 | - | 11 - 12 | 3069 | 133 | ||
BC037674.1 | - | 5 - 6 | 616 | 133 | ||
BC139434.1 | - | 11 - 12 | 2824 | 8862456 | 133 | |
BC145625.1 | - | 11 - 12 | 2806 | 9054193 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |