Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400003398.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003398.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003408.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003408.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003407.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003407.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003409.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003409.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003400.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003400.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003406.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003406.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003411.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003411.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003401.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003401.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003404.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003404.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003399.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003399.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003402.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003402.mm.1 |
TC1400000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003405.mm.1 |
NM_001004436.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003405.mm.1 |
TC1400000381.mm.1 |
MTA TC |