Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014968.1 | - | 5 - 6 | 614 | 96 | |
AK035181.1 | - | 6 - 7 | 1129 | 96 | ||
BC115878.1 | - | 5 - 6 | 938 | 40088737 | 96 | |
BC116658.1 | - | 5 - 6 | 938 | 40088738 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |