Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167967.2 | NP_001161439.1 | 1 - 2 | 348 | 56 | |
NM_001281974.1 | NP_001268903.1 | 1 - 2 | 348 | 56 | ||
NM_001281975.1 | NP_001268904.1 | 1 - 2 | 348 | 56 | ||
NM_133971.3 | NP_598732.2 | 1 - 2 | 348 | 56 | ||
GenBank mRNA | AK031808.1 | - | 1 - 2 | 346 | 56 | |
AK033623.1 | - | 1 - 2 | 346 | 56 | ||
AK038156.1 | - | 1 - 2 | 346 | 56 | ||
AK050983.1 | - | 1 - 2 | 349 | 56 | ||
AK081754.1 | - | 1 - 2 | 346 | 56 | ||
AK082875.1 | - | 1 - 2 | 345 | 56 | ||
AK084456.1 | - | 1 - 2 | 346 | 56 | ||
AK088371.1 | - | 1 - 2 | 298 | 56 | ||
AK143253.1 | - | 1 - 2 | 346 | 56 | ||
BC002198.1 | - | 1 - 2 | 280 | 3489133 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |