Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294138.1 | NP_001281067.1 | 18 - 19 | 1884 | 68 | |
NM_001294140.1 | NP_001281069.1 | 18 - 19 | 1818 | 68 | ||
NM_008194.3 | NP_032220.1 | 17 - 18 | 1800 | 68 | ||
NM_212444.2 | NP_997609.1 | 17 - 18 | 1800 | 68 | ||
GenBank mRNA | AK044308.1 | - | 18 - 19 | 1656 | 68 | |
AK088373.1 | - | 17 - 18 | 1619 | 68 | ||
AK169424.1 | - | 18 - 19 | 1690 | 68 | ||
AK169525.1 | - | 17 - 18 | 1801 | 68 | ||
BC003767.1 | - | 17 - 18 | 1603 | 3593896 | 68 | |
CF740682.1 | - | 2 - 3 | 279 | 68 | ||
U48403.1 | - | 17 - 18 | 1577 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |