Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00005526.mm.1 |
NM_025729.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005526.mm.1 |
NM_025729.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005526.mm.1 |
TC0X00000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005529.mm.1 |
NM_025729.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005529.mm.1 |
NM_025729.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005529.mm.1 |
TC0X00000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005516.mm.1 |
NM_025729.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005516.mm.1 |
NM_025729.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005516.mm.1 |
TC0X00000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005521.mm.1 |
NM_025729.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005521.mm.1 |
NM_025729.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005521.mm.1 |
TC0X00000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005528.mm.1 |
NM_025729.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005528.mm.1 |
NM_025729.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005528.mm.1 |
TC0X00000832.mm.1 |
MTA TC |