Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010084.2 | NP_034214.2 | 11 - 12 | 1094 | 133 | |
GenBank mRNA | AF167405.1 | - | 11 - 12 | 1092 | 133 | |
AK076633.1 | - | 11 - 12 | 1098 | 133 | ||
BC051136.1 | - | 11 - 12 | 1071 | 6708158 | 133 | |
BC144841.1 | - | 11 - 12 | 1067 | 133 | ||
BC145751.1 | - | 11 - 12 | 1067 | 40131054 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |