Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153115.1 | NP_694755.1 | 1 - 2 | 110 | 73 | |
GenBank mRNA | AB089183.1 | - | 1 - 2 | 110 | 73 | |
AF384173.1 | - | 1 - 2 | 73 | 73 | ||
AK020333.1 | - | 1 - 2 | 114 | 73 | ||
BC089530.1 | - | 1 - 2 | 110 | 73 | ||
BC120808.1 | - | 1 - 2 | 75 | 40129731 | 73 | |
BC120810.1 | - | 1 - 2 | 75 | 40129733 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |