Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800000865.mm.1 |
NM_009233.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000865.mm.1 |
NM_009233.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000865.mm.1 |
TC0800000105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800000862.mm.1 |
NM_009233.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000862.mm.1 |
NM_009233.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000862.mm.1 |
TC0800000105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800000864.mm.1 |
NM_009233.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000864.mm.1 |
NM_009233.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000864.mm.1 |
TC0800000105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800000863.mm.1 |
NM_009233.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000863.mm.1 |
NM_009233.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000863.mm.1 |
TC0800000105.mm.1 |
MTA TC |