Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800015213.mm.1 |
NM_177089.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015213.mm.1 |
NM_199323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015213.mm.1 |
TC0800001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800015216.mm.1 |
NM_177089.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015216.mm.1 |
TC0800001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800015214.mm.1 |
NM_177089.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015214.mm.1 |
NM_199323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015214.mm.1 |
TC0800001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800015219.mm.1 |
NM_177089.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015219.mm.1 |
TC0800001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800015217.mm.1 |
NM_177089.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800015217.mm.1 |
TC0800001906.mm.1 |
MTA TC |