Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011960.2 | NP_036090.2 | 9 - 10 | 2242 | 86 | |
GenBank mRNA | AF079557.1 | - | 9 - 10 | 1991 | 86 | |
AK034542.1 | - | 2 - 3 | 174 | 86 | ||
AK036656.1 | - | 9 - 10 | 2265 | 86 | ||
AK088708.1 | - | 6 - 7 | 597 | 86 | ||
AK164013.1 | - | 9 - 10 | 2229 | 86 | ||
BC050892.1 | - | 9 - 10 | 2241 | 86 | ||
BC053037.1 | - | 9 - 10 | 2225 | 86 | ||
BC059827.1 | - | 9 - 10 | 2251 | 86 | ||
EF382673.1 | - | 7 - 8 | 951 | 86 | ||
JQ890227.1 | - | 6 - 7 | 748 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |