Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153196.1 | NP_694876.1 | 3 - 4 | 296 | 93 | |
GenBank mRNA | AK017229.1 | - | 3 - 4 | 314 | 93 | |
AK138939.1 | - | 3 - 4 | 314 | 93 | ||
BC023339.1 | - | 3 - 4 | 296 | 5253827 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |