Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081137.1 | NP_001074606.1 | 20 - 21 | 2400 | 78 | |
GenBank mRNA | EU937530.1 | - | 21 - 22 | 2463 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |