Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133786.3 | NP_598547.1 | 16 - 17 | 2597 | 125 | |
GenBank mRNA | AJ534940.2 | - | 15 - 16 | 2473 | 125 | |
AK048650.1 | - | 11 - 12 | 1811 | 125 | ||
BC061481.1 | - | 16 - 17 | 2572 | 6512832 | 125 | |
BC062939.1 | - | 16 - 17 | 2597 | 6841276 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |