Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035239.2 | NP_001030316.1 | 6 - 7 | 978 | 80 | |
NM_001035240.2 | NP_001030317.1 | 6 - 7 | 978 | 80 | ||
NM_001035241.2 | NP_001030318.1 | 6 - 7 | 978 | 80 | ||
NM_001035242.1 | NP_001030319.1 | 6 - 7 | 978 | 80 | ||
NM_001035243.2 | NP_001030320.1 | 6 - 7 | 978 | 80 | ||
NM_001035245.1 | NP_001030322.1 | 6 - 7 | 1026 | 80 | ||
NM_177341.4 | NP_796315.2 | 6 - 7 | 1045 | 80 | ||
GenBank mRNA | AJ544532.1 | - | 6 - 7 | 978 | 80 | |
AJ544533.1 | - | 6 - 7 | 978 | 80 | ||
AJ544534.1 | - | 6 - 7 | 978 | 80 | ||
AJ544535.1 | - | 6 - 7 | 978 | 80 | ||
AJ544536.1 | - | 6 - 7 | 978 | 80 | ||
AK044454.1 | - | 6 - 7 | 1047 | 80 | ||
AK053673.1 | - | 6 - 7 | 863 | 80 | ||
AK082141.1 | - | 6 - 7 | 860 | 80 | ||
AK173218.1 | - | 6 - 7 | 1049 | 80 | ||
BC037449.1 | - | 6 - 7 | 874 | 80 | ||
BC141400.1 | - | 6 - 7 | 1021 | 9056013 | 80 | |
BC141412.1 | - | 6 - 7 | 1021 | 9056025 | 80 | |
HM064407.1 | - | 6 - 7 | 1070 | 80 | ||
HM064408.1 | - | 6 - 7 | 1070 | 80 | ||
HM064409.1 | - | 6 - 7 | 1070 | 80 | ||
HM064411.1 | - | 6 - 7 | 1070 | 80 | ||
JF706722.1 | - | 6 - 7 | 1041 | 80 | ||
JX644970.1 | - | 6 - 7 | 1070 | 80 | ||
JX644971.1 | - | 6 - 7 | 1070 | 80 | ||
JX644972.1 | - | 6 - 7 | 1070 | 80 | ||
JX644973.1 | - | 6 - 7 | 1070 | 80 | ||
JX644974.1 | - | 6 - 7 | 1070 | 80 | ||
JX644976.1 | - | 6 - 7 | 1070 | 80 | ||
JX644977.1 | - | 6 - 7 | 1070 | 80 | ||
JX644978.1 | - | 6 - 7 | 1070 | 80 | ||
JX644983.1 | - | 6 - 7 | 1070 | 80 | ||
JX644984.1 | - | 6 - 7 | 1070 | 80 | ||
JX644985.1 | - | 6 - 7 | 1070 | 80 | ||
JX644986.1 | - | 6 - 7 | 1070 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |