Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145221.2 | NP_660256.1 | 4 - 5 | 378 | 70 | |
GenBank mRNA | AK013715.1 | - | 4 - 5 | 375 | 70 | |
AK030445.1 | - | 4 - 5 | 258 | 70 | ||
AK033566.1 | - | 4 - 5 | 349 | 70 | ||
AK045438.1 | - | 4 - 5 | 314 | 70 | ||
AK049307.1 | - | 4 - 5 | 365 | 70 | ||
AK088080.1 | - | 4 - 5 | 329 | 70 | ||
AK146356.1 | - | 4 - 5 | 349 | 70 | ||
AY113705.1 | - | 4 - 5 | 343 | 70 | ||
BC063751.1 | - | 4 - 5 | 390 | 6511401 | 70 | |
BU514763.1 | - | 4 - 5 | 389 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |